1-Benzyl-3,5-dibromo-1H-1,2,4-triazole 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR9607-1G OR9607-5G
77412-228EA 52.98 USD
77412-228 77412-230
1-Benzyl-3,5-dibromo-1H-1,2,4-triazole 95%
1-Benzyl-3,5-dibromo-1H-1,2,4-triazole
Formula: C₉H₇Br₂N₃
Melting Pt: 65…67 °C
Storage Temperature: Ambient
MDL Number: MFCD01568839
CAS Number: 106724-85-0
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR