4H,5H,6H,7H,8H-Thieno[3,2-b]azepin-5-one 95%

Supplier: APOLLO SCIENTIFIC

OR931890-1G OR931890-250MG
77411-988EA 338.24 USD
77411-988 77411-990
4H,5H,6H,7H,8H-Thieno[3,2-b]azepin-5-one 95%
4H,5H,6H,7H,8H-Thieno[3,2-b]azepin-5-one
Formula: C₈H₉NOS
Storage Temperature: Ambient
MDL Number: MFCD00980799
CAS Number: 4751-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR