1-Chloro-3-methoxypropane 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: 3-Chloropropyl methyll ether

OR906289-100G OR906289-25G
77411-564EA 475.78 USD
77411-564 77411-566
1-Chloro-3-methoxypropane 98%
1-Chloro-3-methoxypropane
Formula: C₄H₉ClO
MW: 108.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663419
CAS Number: 36215-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR