N-Methyl-p-toluidine 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: N,4-Dimethylaniline, p-(Methylamino)toluene, N-Methyl-4-methylaniline, 4-(Methylamino)toluene, 4-Methyl-N-methylaniline

OR905934-100G OR905934-25G
77411-556EA 41.78 USD
77411-556 77411-558
N-Methyl-p-toluidine 98%
N-Methyl-p-toluidine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 89
Storage Temperature: Ambient
MDL Number: MFCD00025628
CAS Number: 623-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR