1-Butylpyrazole 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: 1-Butyl-1H-pyrazole

OR959882-1G OR959882-5G
77412-208EA 71.8 USD
77412-208 77412-210 77412-212
1-Butylpyrazole 98%
1-Butylpyrazole
Formula: C₇H₁₂N₂
MW: 124.184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08701137
CAS Number: 52096-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR