2-Chloro-5-cyanobenzene-1-sulfonyl chloride 95%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR931550-1G
OR931550-250MG
OR931550-5G
77411-972EA
101.88
USD
77411-972
77411-974
77411-976
2-Chloro-5-cyanobenzene-1-sulfonyl chloride 95%
2-Chloro-5-cyanobenzene-1-sulfonyl chloride
Formula:
C₇H₃Cl₂NO₂S Storage Temperature: Refrigerator |
MDL Number:
MFCD18392427 CAS Number: 942199-56-6 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...