1-Vinyl-2-pyrrolidone 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: N-Vinyl-2-pyrrolidone, 1-Vinyl-2-pyrrolidinone, N-Vinyl-2-pyrrolidinone

OR924885-100G OR924885-25G OR924885-5G
77411-884EA 68.26 USD
77411-884 77411-886 77411-888
1-Vinyl-2-pyrrolidone 98%
1-Vinyl-2-pyrrolidone
Formula: C₆H₉NO
MW: 111.1418 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00003197
CAS Number: 88-12-0
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR