Phenylacetylene

Supplier: APOLLO SCIENTIFIC
Danger

OR8217-100G OR8217-25G OR8217-500G
77411-368EA 59.1 USD
77411-368 77411-370 77411-372
Phenylacetylene
Phenylacetylene
Formula: C₈H₆
Boiling Pt: 142…144 °C
Melting Pt: –45 °C
Density: 0.93 g/cm³ (25 °C)
Flash Pt: 27 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008570
CAS Number: 536-74-3
EINECS: 208-645-1
UN: 3295
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR