(Naphth-2-yl)acetyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

OR8139-10G OR8139-1G OR8139-250MG
77411-356EA 358.62 USD
77411-356 77411-358 77411-360
(Naphth-2-yl)acetyl chloride
(Naphth-2-yl)acetyl chloride
Formula: C₁₂H₉ClO
Melting Pt: 60…61 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02180524
CAS Number: 37859-25-9
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR