Aluminium bromide, anhydrous

Supplier: APOLLO SCIENTIFIC
Danger

OR7306-100G OR7306-25G
77411-204EA 178.02 USD
77411-204 77411-206
Aluminium bromide, anhydrous
Aluminium bromide
Formula: AlBr₃
Boiling Pt: 265 °C
Melting Pt: 97 °C
Density: 3.205 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003421
CAS Number: 7727-15-3
EINECS: 231-779-7
UN: 1725
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR