Copper(I)-2-thiophenecarboxylate 95%

Supplier: APOLLO SCIENTIFIC

Synonyms: (2-Thiophenecarboxylato)copper(I), Copper(I) thiophene-2-carboxylate, 2-Thiophenecarboxylic Acid Copper(I) Salt, CuTC

OR936291-100G OR936291-25G
77412-002EA 217 USD
77412-002 77412-004
Copper(I)-2-thiophenecarboxylate 95%
Copper(I)-2-thiophenecarboxylate
Formula: C₅H₃CuO₂S
Storage Temperature: Ambient
MDL Number: MFCD02183524
CAS Number: 68986-76-5
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR