(S)-(-)-2-Methoxypropionic acid 97%
Supplier: APOLLO SCIENTIFIC
Synonyms:
(S)-2-Methoxypropanoic acid
OR923407-1G
OR923407-25G
OR923407-5G
77411-846EA
32.6
USD
77411-846
77411-848
77411-850
(S)-(-)-2-Methoxypropionic acid 97%
(S)-(-)-2-Methoxypropionic acid
Formula:
C₄H₈O₃ MW: 104.105 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01632589 CAS Number: 23953-00-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...