1-Chloro-2-methylpentane 98%

Supplier: APOLLO SCIENTIFIC

OR923251-1G OR923251-25G OR923251-5G
77411-834EA 41.78 USD
77411-834 77411-836 77411-838
1-Chloro-2-methylpentane 98%
1-Chloro-2-methylpentane
Formula: C₆H₁₃Cl
Storage Temperature: Ambient
MDL Number: MFCD00053236
CAS Number: 14753-05-0
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR