N-Methyl-4-(pyridin-2-yl)benzylamine

Supplier: APOLLO SCIENTIFIC
Danger

OR9134-1G OR9134-250MG
77411-684EA 309.72 USD
77411-684 77411-686
N-Methyl-4-(pyridin-2-yl)benzylamine
N-Methyl-4-(pyridin-2-yl)benzylamine
Formula: C₁₃H₁₄N₂
Storage Temperature: Ambient
MDL Number: MFCD08271910
CAS Number: 869901-08-6
UN: 2922
ADR: 8+6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR