1-(4-Thiazolyl)ethanone 98%

Supplier: APOLLO SCIENTIFIC

OR902409-1G OR902409-250MG OR902409-5G
77411-508EA 110.04 USD
77411-508 77411-510 77411-512
1-(4-Thiazolyl)ethanone 98%
1-(4-Thiazolyl)ethanone
Formula: C₅H₅NOS
MW: 127.1643 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03550493
CAS Number: 38205-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR