1-Methyl-1H-indole-4-sulfonyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR8646-1G OR8646-250MG
77411-430EA 467.64 USD
77411-430 77411-432
1-Methyl-1H-indole-4-sulfonyl chloride 97%
1-Methyl-1H-indole-4-sulfonyl chloride
Formula: C₉H₈ClNO₂S
Melting Pt: 95…97 °C
Storage Temperature: Ambient
MDL Number: MFCD08690260
CAS Number: 876316-36-8
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR