2-(Ethylthio)ethylamine 96%

Supplier: APOLLO SCIENTIFIC
Danger

OR8640-25G OR8640-5G
77411-420EA 320.92 USD
77411-420 77411-422
2-(Ethylthio)ethylamine 96%
2-(Ethylthio)ethylamine
Formula: C₄H₁₁NS
Boiling Pt: 159…160 °C
Density: 0.976 g/cm³ (25 °C)
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00014826
CAS Number: 36489-03-9
UN: 2734
ADR: 8+3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR