5-[(Dimethylamino)sulfonyl]-2-methyl-3-furoic acid 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR8125-1G OR8125-250MG
77411-344EA 103.92 USD
77411-344 77411-346
5-[(Dimethylamino)sulfonyl]-2-methyl-3-furoic acid 97%
5-[(Dimethylamino)sulfonyl]-2-methyl-3-furoic acid
Formula: C₈H₁₁NO₅S
MW: 233.24 g/mol
Melting Pt: 169 °C
Storage Temperature: Ambient
MDL Number: MFCD02180761
CAS Number: 306936-39-0
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR