4-Bromo-1-butene 99%

Supplier: APOLLO SCIENTIFIC
Danger

OR8033-100G OR8033-25G OR8033-500G
77411-334EA 59.1 USD
77411-334 77411-336 77411-338
4-Bromo-1-butene 99%
4-Bromo-1-butene
Formula: C₄H₇Br
MW: 135 g/mol
Boiling Pt: 98…100 °C
Density: 1.33 g/cm³ (25 °C)
Flash Pt: 1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000258
CAS Number: 5162-44-7
EINECS: 225-937-4
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR