1-Methyl-1H-indol-5-ylamine 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Amino-1-n-methylindole, 1-Methyl-1H-indol-5-amine, 5-Amino-1-methyl-1H-indole

OR6792-1G OR6792-500MG OR6792-5G
77410-906EA 28.52 USD
77410-906 77410-908 77410-910
1-Methyl-1H-indol-5-ylamine 97%
1-Methyl-1H-indol-5-ylamine
Formula: C₉H₁₀N₂
MW: 146.19 g/mol
Melting Pt: 101…104 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03839873
CAS Number: 102308-97-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR