1-Boc-4-bromopiperidine 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: tert-Butyl 4-Bromopiperidine-1-carboxylate, 4-Bromopiperidine-1-carboxylic acid tert-Butyl ester, 1-Boc-4-bromopiperidine, 1-tert-Butoxycarbonyl-4-bromopiperidine, N-Boc-4-Bromopiperidine

OR962067-1G OR962067-5G
77412-238EA 22.42 USD
77412-238 77412-240
1-Boc-4-bromopiperidine 97%
1-Boc-4-bromopiperidine
Formula: C₁₀H₁₈BrNO₂
MW: 264.1594 g/mol
Boiling Pt: 100…118 °C/0.5 mmHg
Density: 1.3403 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD04115039
CAS Number: 180695-79-8
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR