4-Methoxybutanoic acid 95%

Supplier: APOLLO SCIENTIFIC

OR952214-1G OR952214-250MG
77412-164EA 660.2 USD
77412-164 77412-166
4-Methoxybutanoic acid 95%
4-Methoxybutanoic acid
Formula: C₅H₁₀O₃
MW: 118.131 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00029855
CAS Number: 29006-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR