4-Cyclopropylbenzenesulfonyl chloride 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR950305-1G OR950305-250MG OR950305-5G
77412-154EA 619.44 USD
77412-154 77412-156 77412-158
4-Cyclopropylbenzenesulfonyl chloride 95%
4-Cyclopropylbenzenesulfonyl chloride
Formula: C₉H₉ClO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD09929602
CAS Number: 167404-32-2
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR