p-Tolylhydrazinium chloride 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Methylphenylhydrazinium chloride, 4-Tolylhydrazine hydrochloride, p-Tolylhydrazine hydrochloride, 1-(4-Methylphenyl)hydrazine hydrochloride, 4-Methylphenylhydrazine hydrochloride, 1-(4-Tolyl)hydrazine hydrochloride

OR7543-100G OR7543-500G
77411-238EA 166.06 USD
77411-238 77411-240
p-Tolylhydrazinium chloride 98%
p-Tolylhydrazinium chloride
Formula: C₇H₁₁ClN₂
Melting Pt: 215…218 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00012940
CAS Number: 637-60-5
EINECS: 211-295-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR