3-Iodo-4-methylbenzyl bromide

Supplier: APOLLO SCIENTIFIC
Danger

OR55731-1G OR55731-250MG
77410-518EA 158.94 USD
77410-518 77410-520
3-Iodo-4-methylbenzyl bromide
3-Iodo-4-methylbenzyl bromide
Formula: C₈H₈BrI
Storage Temperature: Refrigerator
MDL Number: MFCD09745120
CAS Number: 626250-55-3
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR