1,5,7-Triazabicyclo[4.4.0]dec-5-ene 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1,3,4,6,7,8-Hexahydro-2H-pyrimido[1,2-a]pyrimidine, Hhpp, 2,3,4,6,7,8-Hexahydro-1H-pyrimido[1,2-a]pyrimidine, TBD

OR52892-100G OR52892-25G OR52892-5G
77410-262EA 470.7 USD
77410-262 77410-264 77410-266
1,5,7-Triazabicyclo[4.4.0]dec-5-ene 97%
1,5,7-Triazabicyclo[4.4.0]dec-5-ene
Formula: C₇H₁₃N₃
MW: 139.20 g/mol
Melting Pt: 125…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00043003
CAS Number: 5807-14-7
UN: 3263
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR