5-Fluoro-2-methylbenzyl bromide 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-(Bromomethyl)-4-fluoro-1-methylbenzene

PC0365-1G PC0365-25G PC0365-5G
77412-518EA 21.32 USD
77412-518 77412-520 77412-522
5-Fluoro-2-methylbenzyl bromide 97%
5-Fluoro-2-methylbenzyl bromide
Formula: C₈H₈BrF
MW: 203.05
Boiling Pt: 84…85 °C/8 mmHg
Flash Pt: 85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01631528
CAS Number: 261951-71-7
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR