5-[5-(Trifluoromethyl)isoxazol-3-yl]thiophene-2-sulfonyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC0042-1G PC0042-5G
77412-350EA 66.22 USD
77412-350 77412-352
5-[5-(Trifluoromethyl)isoxazol-3-yl]thiophene-2-sulfonyl chloride 97%
5-[5-(Trifluoromethyl)isoxazol-3-yl]thiophene-2-sulfonyl chloride
Formula: C₈H₃ClF₃NO₃S₂
Melting Pt: 106…111 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00831001
CAS Number: 229956-98-3
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR