2-Chloro-3,6-difluorobenzylamine 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC0006-1G PC0006-25G PC0006-5G
77412-338EA 41.78 USD
77412-338 77412-340 77412-342
2-Chloro-3,6-difluorobenzylamine 98%
2-Chloro-3,6-difluorobenzylamine
Formula: C₇H₆ClF₂N
Storage Temperature: Ambient
MDL Number: MFCD01631449
CAS Number: 261762-45-2
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR