Diethyl bromomalonate 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR9987-500G OR9987-100G OR9987-25G
77412-334EA 135.5 USD
77412-334 77412-330 77412-332
Diethyl bromomalonate 98%
Diethyl bromomalonate
Formula: C₇H₁₁BrO₄
Boiling Pt: 140…141 °C/30 mmHg
Density: 1.402 g/cm³ (25 °C)
Flash Pt: 111 °C
Storage Temperature: Ambient
MDL Number: MFCD00009138
CAS Number: 685-87-0
EINECS: 211-683-1
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR