4-Bromo-o-toluic acid

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Bromo-2-methylbenzoic acid

OR9537-100G OR9537-25G OR9537-500G
77412-178EA 74.38 USD
77412-178 77412-180 77412-182
4-Bromo-o-toluic acid
4-Bromo-o-toluic acid
Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Melting Pt: 180…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00040905
CAS Number: 68837-59-2
EINECS: 272-437-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR