2-(4-Chloro-2-nitrophenyl)malondialdehyde 95%

Supplier: APOLLO SCIENTIFIC

OR7627-1G OR7627-25G OR7627-5G
77411-268EA 128.38 USD
77411-268 77411-270 77411-272
2-(4-Chloro-2-nitrophenyl)malondialdehyde 95%
2-(4-Chloro-2-nitrophenyl)malondialdehyde
Formula: C₉H₆ClNO₄
Melting Pt: 140…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00216498
CAS Number: 205744-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR