2-tert-Butyl-1H-indole-3-carbaldehyde 95+%

Supplier: APOLLO SCIENTIFIC
Warning

OR7623-1G OR7623-5G
77411-260EA 105.96 USD
77411-260 77411-262
2-tert-Butyl-1H-indole-3-carbaldehyde 95+%
2-tert-Butyl-1H-indole-3-carbaldehyde
Formula: C₁₃H₁₅NO
MW: 201.27 g/mol
Melting Pt: 224 °C
Storage Temperature: Ambient
MDL Number: MFCD01241608
CAS Number: 29957-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR