3-Chloro-2-(morpholin-4-yl)pyridine-4-boronic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Borono-3-chloro-2-(morpholin-4-yl)pyridine, 4-(4-Borono-3-chloropyridin-2-yl)morpholine

OR6094-100MG OR6094-50MG
77410-694EA 124.3 USD
77410-694 77410-696
3-Chloro-2-(morpholin-4-yl)pyridine-4-boronic acid 97%
3-Chloro-2-(morpholin-4-yl)pyridine-4-boronic acid
Formula: C₉H₁₂BClN₂O₃
MW: 242.467 g/mol
Melting Pt: 120…125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09258746
CAS Number: 957061-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR