EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide) 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: EDAC, WSC, EDC, N'-(Ethylcarbonimidoyl)-N,N-dimethylpropane-1,3-diamine, 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, N1-((Ethylimino)methylene)-N3,N3-dimethylpropane-1,3-diamine

OR53094-100G OR53094-25G OR53094-5G
77410-282EA 192.56 USD
77410-282 77410-284 77410-286
EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide) 98%
EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide)
Formula: C₈H₁₇N₃
Density: 0.877 g/cm³ (25 °C)
Storage Temperature: Freezer
MDL Number: MFCD00044916
CAS Number: 1892-57-5
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR