Oxazolo[4,5-b]pyridine 96%

Supplier: APOLLO SCIENTIFIC

OR909642-1G OR909642-5G
77411-596EA 93.74 USD
77411-596 77411-598
Oxazolo[4,5-b]pyridine 96%
Oxazolo[4,5-b]pyridine
Formula: C₆H₄N₂O
MW: 120.109 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13175183
CAS Number: 273-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR