Ethyl acetimidate hydrochloride 95%

Supplier: APOLLO SCIENTIFIC

OR903066-100G OR903066-500G
77411-522EA 99.84 USD
77411-522 77411-524
Ethyl acetimidate hydrochloride 95%
Ethyl acetimidate hydrochloride
Formula: C₄H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD00012572
CAS Number: 2208-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR