2-Oxoindoline-5-sulfonyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR8665-1G OR8665-250MG
77411-436EA 307.68 USD
77411-436 77411-438
2-Oxoindoline-5-sulfonyl chloride 97%
2-Oxoindoline-5-sulfonyl chloride
Formula: C₈H₆ClNO₃S
MW: 231.66 g/mol
Melting Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD06655924
CAS Number: 199328-31-9
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR