4-(Pyrrolidin-3-yl)morpholine

Supplier: APOLLO SCIENTIFIC
Danger

OR54379-1G OR54379-5G
77410-380EA 186.44 USD
77410-380 77410-382
4-(Pyrrolidin-3-yl)morpholine
4-(Pyrrolidin-3-yl)morpholine
Formula: C₈H₁₆N₂O
Storage Temperature: Ambient
MDL Number: MFCD06740352
CAS Number: 53617-37-1
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR