Dichloroacetic acid (DCA) 95%

Supplier: APOLLO SCIENTIFIC

Synonyms: DCA Deblock, DCA

OR922813-100G OR922813-500G
77411-818EA 117.16 USD
77411-818 77411-820
Dichloroacetic acid (DCA) 95%
Dichloroacetic acid (DCA)
Formula: C₂H₂Cl₂O₂
MW: 128.94 g/mol
Boiling Pt: 194
Melting Pt: -4
Storage Temperature: Ambient
MDL Number: MFCD00004223
CAS Number: 79-43-6
ADR: 8 II
Merck Index: 13,03075

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR