(1-Methyl-3-phenyl-1H-pyrazol-5-yl)methylamine 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: [5-(Aminomethyl)-1-methyl-1H-pyrazol-3-yl]benzene, 5-(Aminomethyl)-1-methyl-3-phenyl-1H-pyrazole

OR9209-1G OR9209-250MG
77411-740EA 410.58 USD
77411-740 77411-742
(1-Methyl-3-phenyl-1H-pyrazol-5-yl)methylamine 97%
(1-Methyl-3-phenyl-1H-pyrazol-5-yl)methylamine
Formula: C₁₁H₁₃N₃
MW: 187.241 g/mol
Melting Pt: 47.5…49.5 °C
Storage Temperature: Ambient
MDL Number: MFCD08271943
CAS Number: 876728-39-1
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR