4-Chloro-2-(methylthio)pyrimidine 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Chloro-2-methylthiopyrimidine

OR6414-100G OR6414-25G OR6414-5G
77410-834EA 179.32 USD
77410-834 77410-836 77410-838
4-Chloro-2-(methylthio)pyrimidine 97%
4-Chloro-2-(methylthio)pyrimidine
Formula: C₅H₅ClN₂S
MW: 160.63 g/mol
Boiling Pt: 139…140 °C/36 mmHg
Melting Pt: –2 °C
Density: 1.381 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Freezer
MDL Number: MFCD00006083
CAS Number: 49844-90-8
EINECS: 256-500-6
UN: 1760
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR