Maleic anhydride

Supplier: APOLLO SCIENTIFIC

Synonyms: 2,5-Furanedione

OR61510-100G OR61510-25G
77410-764EA 202.74 USD
77410-764 77410-766
Maleic anhydride
Maleic anhydride
Formula: C₄H₂O₃
Boiling Pt: 202 °C
Melting Pt: 52…56 °C
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00005518
CAS Number: 108-31-6
UN: 2215
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR