1,5-Diazobicyclo[4.3.0]-5-nonene 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR5394-100G
OR5394-1KG
OR5394-250G
77410-338EA
96.78
USD
77410-338
77410-340
77410-342
1,5-Diazobicyclo[4.3.0]-5-nonene 98%
1,5-Diazobicyclo[4.3.0]-5-nonene
Formula:
C₇H₁₂N₂ Boiling Pt: 95…98 °C/7.5 mmHg Density: 1.005 g/cm³ (25 °C) Flash Pt: 94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005554 CAS Number: 3001-72-7 EINECS: 221-087-3 UN: 3267 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...