Benzhydrylamine

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: α-Phenylbenzylamine, α-Aminodiphenylmethane, Diphenylmethanamine, Aminodiphenylmethane, 1,1-Diphenylmethanamine, Benzhydrylamin

OR8244-100G OR8244-1G OR8244-25G
77411-384EA 103.92 USD
77411-384 77411-386 77411-388
Benzhydrylamine
Benzhydrylamine
Formula: C₁₃H₁₃N
Boiling Pt: 294…296 °C
Melting Pt: 41579 °C
Density: 1.063 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008059
CAS Number: 91-00-9
EINECS: 202-032-2
UN: 2735
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR