[(4-Bromophenyl)sulfonyl]acetonitrile

Supplier: APOLLO SCIENTIFIC
Warning

OR7778-1G OR7778-25G OR7778-5G
77411-290EA 48.9 USD
77411-290 77411-292 77411-294
[(4-Bromophenyl)sulfonyl]acetonitrile
[(4-Bromophenyl)sulfonyl]acetonitrile
Formula: C₈H₆BrNO₂S
Melting Pt: 190…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00129455
CAS Number: 126891-45-0
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR