4-Thiazolecarbonyl chloride 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR6374-1G OR6374-250MG
77410-824EA 187.46 USD
77410-824 77410-826
4-Thiazolecarbonyl chloride 95%
4-Thiazolecarbonyl chloride
Formula: C₄H₂ClNOS
Storage Temperature: Refrigerator
CAS Number: 3745-79-7
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR