2-Bromo-1-(tetrahydro-2H-pyran-4-yl)ethanone 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR55638-1G OR55638-250MG OR55638-5G
77410-482EA 131.42 USD
77410-482 77410-484 77410-486
2-Bromo-1-(tetrahydro-2H-pyran-4-yl)ethanone 95%
2-Bromo-1-(tetrahydro-2H-pyran-4-yl)ethanone
Formula: C₇H₁₁BrO₂
MW: 207.065 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11656452
CAS Number: 141095-78-5
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR