4-Ethynyltetrahydro-2H-pyran 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR55577-1G OR55577-250MG OR55577-5G
77410-474EA 217 USD
77410-474 77410-476 77410-478
4-Ethynyltetrahydro-2H-pyran 97%
4-Ethynyltetrahydro-2H-pyran
Formula: C₇H₁₀O
Storage Temperature: Refrigerator
MDL Number: MFCD17016081
CAS Number: 1202245-65-5
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR