4-(Cyclopropyl)benzyl bromide

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-(Bromomethyl)-4-cyclopropylbenzene

OR54647-1G OR54647-250MG OR54647-5G
77410-402EA 344.36 USD
77410-402 77410-404 77410-406
4-(Cyclopropyl)benzyl bromide
4-(Cyclopropyl)benzyl bromide
Formula: C₁₀H₁₁Br
MW: 211.098 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12024391
CAS Number: 1150617-57-4
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR